VWR®, Vanadium standard solutions for ICP-MS, ARISTAR®
Leverandør: VWR Chemicals
456972BEA 1925 NOK
456972B 85643.180
VWR®, Vanadium standard solutions for ICP-MS, ARISTAR®
Standarder Kationstandarder Vanadium Standards
  • Produced from high purity acids, water ASTM I 18 Megaohms
  • Solution assayed by titration
  • Final concentration verified against reference material from NIST
  • Total maximum uncertainty: 0,5% (typical)
  • Delivered with complete certificate of analysis
Sertifiseringer: Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities
Limited Time Sale
Select items below are currently on sale