Wolfram 10.000 mg/l i vann (fra W) ARISTAR® enkeltgrunnstoffsstandard til ICP
Leverandør: VWR Chemicals
457172EEA 5550 NOK
457172E 457174G
Wolfram 10.000 mg/l i vann (fra W) ARISTAR® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Tungsten Standards
W in H₂O tr.NH₄OH.
Traceable to SRM from NIST, tested in an ISO Guide 34 / ISO17025 accredited laboratory.
Supplied with certificate of analysis.