VWR® ARISTAR®, Boron Standard Solutions for ICP-MS
Leverandør: VWR Chemicals
85551.180EA 960 NOK
85551.180 457213B
VWR® ARISTAR®, Boron Standard Solutions for ICP-MS
Standarder Kationstandarder Boron Standards
  • Produced from high purity acids, water ASTM I 18 Megaohms
  • Solution assayed by titration
  • Final concentration verified against reference material from NIST
  • Total maximum uncertainty: 0,5% (typical)
  • Delivered with complete Certificate of Analysis
Sertifiseringer: Traceable to SRM from NIST. Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.