Supelco® Organic Reference Standard, 2-Methylnaphthalene

Leverandør: Merck

45796-250MG
45796-250MGEA 685 NOK
45796-250MG
Supelco® Organic Reference Standard, 2-Methylnaphthalene
Standarder Organiske standarder

  • GHS signal: GHS07, GHS09


NB: P261 - P264 - P273 - P301 + P312 - P302 + P352 - P305 + P351 + P338

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45400eea6c42600a0c646d73d65c39acf7b6aff7&vid=45400eea6c42600a0c646d73d65c39acf7b6aff7&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Synonym(s) β-Methylnaphthalene
Environmentally Preferable
M-Clarity quality level MQ100
CAS-nr 91-57-6
MDL No. MFCD00004118
Molekylvekt 142,20
Hardhetsklasse beskrivelse H302 - H315 - H319 - H335 - H411
Fraktforhold STD
Lagringsforhold 2...30 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR