Klorid 200 mg/l i vann (fra NaCl) ARISTAR® enkeltgrunnstoffsstandard for ionekromatografi
Leverandør: VWR Chemicals
458082HEA 875 NOK
458082H
Klorid 200 mg/l i vann (fra NaCl) ARISTAR® enkeltgrunnstoffsstandard for ionekromatografi
Standarder Anionstandarder Chloride Standards
(Cl in H₂O).
Traceable to SRM from NIST, tested in an ISO Guide 34 / ISO17025 accredited laboratory
Supplied with certificate of analysis.