VWR® ARISTAR® Anion Multi-Component Standard 1 for Ion Chromatography, Avantor
Leverandør: VWR Chemicals
458142CEA 4250 NOK
458142C
VWR® ARISTAR® Anion Multi-Component Standard 1 for Ion Chromatography, Avantor
Standarder Anionstandarder Multi-Element Anioinic Standards
Cl 30 ppm   PO150 ppm
F 20 ppm   SO150 ppm
NO100 ppm
Traceable to SRM from NIST, tested in an ISO Guide 34 / ISO17025 accredited laboratory
Supplied with certificate of analysis.