VWR® ARISTAR® Multi-Component Anion Mix 9, Avantor
Leverandør: VWR Chemicals
458164CEA 3625 NOK
458164C
VWR® ARISTAR® Multi-Component Anion Mix 9, Avantor
Standarder Anionstandarder Multi-Element Anioinic Standards
Two solutions - a nitrite standard solution and a 6 component multi-element standard solution.
Br 40 ppm   NO3 40 ppm
Cl 40 ppm   PO4 40 ppm
F 20 ppm   SO4 40 ppm
NO2 40 ppm
Traceable to SRM from NIST, tested in an ISO Guide 34 / ISO17025 accredited laboratory
Supplied with certificate of analysis.