VWR® ARISTAR® Multi-Component Anion Mix 10, Avantor
Leverandør: VWR Chemicals
458174AEA 3150 NOK
458174A
VWR® ARISTAR® Multi-Component Anion Mix 10, Avantor
Standarder Anionstandarder Multi-Element Anioinic Standards
Two solutions - a nitrite standard solution and also a 6 component multi-element standard solution.
Br 50 ppm   NO3 50 ppm
Cl 50 ppm   PO4 50 ppm
F 25 ppm   SO4 50 ppm
NO2 50 ppm
Traceable to SRM from NIST, tested in an ISO Guide 34 / ISO17025 accredited laboratory.
Supplied with certificate of analysis.