Kalsium 1.000 mg/l i fortynnet salpetersyre (fra Ca) ARISTAR® enkeltgrunnstoffsstandard til ionekromatografi
Leverandør: VWR Chemicals
458202UEA 1375 NOK
458202U
Kalsium 1.000 mg/l i fortynnet salpetersyre (fra Ca) ARISTAR® enkeltgrunnstoffsstandard til ionekromatografi
Standarder Kationstandarder Calcium Standards
Ca in dilute HNO₃.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.