Magnesium 1.000 mg/l i fortynnet salpetersyre (fra Mg) ARISTAR® enkeltgrunnstoffsstandard til ionekromatografi
Leverandør: VWR Chemicals
458212WEA 1375 NOK
458212W
Magnesium 1.000 mg/l i fortynnet salpetersyre (fra Mg) ARISTAR® enkeltgrunnstoffsstandard til ionekromatografi
Standarder Kationstandarder Magnesium Standards
Mg in H₂O tr.HNO₃.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Sertifiseringer: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Leveringsinformasjon: Supplied with certificate of analysis.