Litium 1.000 mg/l, ARISTAR® enkeltgrunnstoffsstandard til ionekromatografi
Leverandør: VWR Chemicals
458242FEA 1375 NOK
458242F
Litium 1.000 mg/l, ARISTAR® enkeltgrunnstoffsstandard til ionekromatografi
Standarder Kationstandarder Lithium Standards
Li in H₂O tr.HNO₃.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.