Litium 200 mg/l i fortynnet salpetersyre (fra Li) ARISTAR® enkeltgrunnstoffsstandard til ionekromatografi
Leverandør: VWR Chemicals
458322EEA 1150 NOK
458322E
Litium 200 mg/l i fortynnet salpetersyre (fra Li) ARISTAR® enkeltgrunnstoffsstandard til ionekromatografi
Standarder Kationstandarder Lithium Standards
Li in H₂O tr.HNO₃.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.