VWR®, Combustion Boats
Leverandør: VWR Collection
459-0223EA 1050 NOK
459-0223 459-0226 459-0224 459-0225
VWR®, Combustion Boats
Skip Forbrenningsskip
Porcelain.
  • Glazed boats can withstand temperatures up to 1000 °C, unglazed up to 1350 °C
  • Eyelet to aid recovery from furnace
  • Glazed porcelain has high resistance to acids and bases