VWR®, Incinerating Dishes
Leverandør: VWR Collection
459-0233EA 535 NOK
459-0233 459-0232 459-0231 459-0230 459-0229 459-0228 459-0227
VWR®, Incinerating Dishes
Skip Forbrenningsskip
Porcelain, glazed, produced according to DIN 12904.
  • Dishes can withstand temperatures up to 1000 °C
  • Stable with large flat unglazed base
  • Glazed porcelain has high resistance to acids and bases