Forbrenningsskål
Leverandør: MORGAN ADVANCED MATERIALS HALD
L33/2NUMBAB100
459-2228EA 0 NOK
459-2228
Forbrenningsskål
Skip Forbrenningsskip
Porselen.
  • Glassert på inn- og utsiden unntatt den ytre bunnoverflaten
  • "God kjemikaliebestandighet
  • Varmebestandighet opptil 1000 °C"
Numbers 1 to 99 or 100 to 999 are printed prior to glazing so that the identification of dishes is simple and not affected by the high temperatures of incineration.
Bestillingsinformasjon: At time of order, please request the identification number required.