VWR®, digler, rustfritt stål 304
Leverandør: VWR Collection
459-4501EA 290 NOK
459-4501 459-4502 459-4503
VWR®, digler, rustfritt stål 304
Digler
These crucibles are made from stainless steel 304.
  • Highly resistant to chemicals and reagents
  • Withstand maximum temperature of 800 °C
Without lid.