Isopropylacetat ≥99.5% (by GC), Puriss. p.a., Riedel-de Haën™

Leverandør: Honeywell Chemicals
Danger

45960-1L 45960-250ML
45960-1LEA 1200 NOK
45960-1L 45960-250ML
Isopropylacetat ≥99.5% (by GC), Puriss. p.a., Riedel-de Haën™
Isopropylacetat

Beilstein Number: 1740761

Formel: C₅H₁₀O₂
Kokepunkt: 85 °C
Smeltepunkt: –62 °C
Tetthet: 0,8662 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008877
CAS-nummer: 108-21-4
EINECS: 203-561-1
FN: 1220
ADR: 3,II
Merck Indeks: 13,05225

Bestill nå

Spesifikasjon Testresultater

assay (GC) Min. 99 %
measurement uncertainty effective %
identity (NMR) complying
refractive index (n 25/D) 1.38 - 1.39
water (Karl Fischer) Max. 0.5 %
Aluminium (Al) Max. 0.5 ppm
Barium (Ba) Max. 0.1 ppm
Bismuth (Bi) Max. 0.1 ppm
Calcium (Ca) Max. 0.5 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.02 ppm
Chromium (Cr) Max. 0.02 ppm
Copper (Cu) Max. 0.02 ppm
Iron (Fe) Max. 0.1 ppm
Potassium (K) Max. 0.5 ppm
Lithium (Li) Max. 0.1 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.02 ppm
Molybdenum (Mo) Max. 0.1 ppm
Sodium (Na) Max. 0.5 ppm
Nickel (Ni) Max. 0.02 ppm
Lead (Pb) Max. 0.1 ppm
Strontium (Sr) Max. 0.1 ppm
Zinc (Zn) Max. 0.1 ppm
Autoignition Temperature 894 °F


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR