Okadaic acid sodium salt, Millipore®

Leverandør: Merck Millipore Calbiochem

459620-25
459620-25EA 2400 NOK
459620-25
Okadaic acid sodium salt, Millipore®
Okadaic acid sodium salt
Formel: C₄₄H₆₇NaO₁₃
MDL nummer: MFCD00210208
CAS-nummer: 209266-80-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR