Vann, Molecular Biology Grade, meets analytical specification of USP Molecular biology grade, sterile, Corning®

Leverandør: Corning

Synonymer: Hydrogen oxide

46-000-CM 46-000-CI 46-000-CV
46-000-CMEA 2250 NOK
46-000-CM MDTC46-000-CI MDTC46-000-CV
Vann, Molecular Biology Grade, meets analytical specification of USP Molecular biology grade, sterile, Corning®
Vann

Molecular biology grade water is ideal for a wide variety of routine tasks requiring a high degree of sterility Molecular biology grade water is ideal for the preparation of reagents, rinsing glassware and plasticware, and other molecular biology applications where RNase-, DNase-, and Protease-free water is required. No toxic agents, including DEPC, are used in the preparation of Mediatech's molecular biology grade water, eliminating possible interferences with enzymatic reactions.


Sertifiseringer: Molecular Biology Grade Water Tested to USP Sterile Purified Water Specifications

Formel: H₂O
MW: 18.02 g/mol
Kokepunkt: 100
Smeltepunkt: 0
MDL nummer: MFCD00011332
CAS-nummer: 7732-18-5
Merck Indeks: 13,10098

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR