Phosphate-Buffered saline (PBS), 10X concentrate, pH 7,4, molecular biology grade, Corning®
Leverandør: Corning
46-013-CM
46-013-CMEA 5625 NOK
46-013-CM
Phosphate-Buffered saline (PBS), 10X concentrate, pH 7,4, molecular biology grade, Corning®
Buffere Buffere, cellekultur
Phosphate Buffered Saline (PBS) is used to maintain a physiological pH and osmolarity, two key factors in tissue culture and molecular biology research
  • Manufactured in a cGMP environment
  • Formulated with high quality WFI water
  • RNase -, DNase-, and Protease-free
  • PBS without calcium and magnesium
  • pH 7.4 ±0.1
Phosphate Buffered Saline is used to maintain a physiological pH and osmolarity, two key factors in tissue culture and molecular biology research. This concentrated form should be diluted to a working concentration using reagent grade water.
NB: For research use only.