VWR® Thermal Shake lite, Thermal Shaker
Leverandør: VWR Collection
460-0249EA 25075 NOK
460-0249
VWR® Thermal Shake lite, Thermal Shaker
Termostatblokker Termostatblokker med ristefunksjon
The combination of heating/cooling and shaking makes the Thermal Shake lite microtube shaking incubator ideal for many life science research applications in molecular biology, biochemistry and clinical chemistry. Its compact footprint incorporates an intuitive control panel with large multicolour display, allowing users to easily program and view temperature, time and speed settings.
  • Choice of eight interchangeable aluminium blocks accommodate PCR plates and tubes ranging from 0,2 to 15 ml
  • Fine tune speed control
  • Rapid heating and cooling
  • Compact footprint
More Product Information
Bestill nå
SPESIFIKASJONER
Modell Thermal Shake lite
Environmentally Preferable
kretsløp 3 mm
Temperaturkontrollområde 14 below ambient to 100 °C
Oppvarmingshastighet 6,5 °C/min
Temperaturnøyakighet ±0,5 °C
B×D×H 330×166×240 mm
Vekt 8,5 kg
Hastighetsområde 300 - 1500 min⁻¹