Digital block heater, compact
Leverandør: Thermo Fisher Scientific
88871003 88871004
460-0284EA 4600 NOK
460-0284 460-0283
Digital block heater, compact
Termostatblokker Digitale termostatblokker
These versatile digital block heaters provide good temperature uniformity and stability ensuring reproducible results. Units are compact, light weight with digital temperature controls and display, they maximises valuable bench space. The units are ideal for sample preparation, enzyme preservation, DNA amplification and enzyme–substrate reactions.
  • Built-in temperature-sensing probe assists with outstanding temperature accuracy and control
  • Designed for precise temperature control
  • Fast heating rate
  • Multiple combination of different size of blocks allows
More Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Temperaturområde RT* +2...100 °C
Temperaturstabilitet ≤±0,5 °C