VWR®, Digital Mini Block Heaters

Leverandør: VWR Collection
460-0334EA 7300 NOK
460-0334 460-0335 460-0336 460-0337
VWR®, Digital Mini Block Heaters
Termostatblokker Digitale termostatblokker
These mini block heaters are ideal for applications that require accurate, repeatable results. These personal-sized units take up minimal bench space. Choice of model with or without heated lid. Units are ideal for immunoassays, ELISA, LAMP assays, enzyme reactions, denaturations and blood banking.

  • LED display for temperature and time
  • Temperature range from ambient 5 to 100 °C
  • Temperature accuracy: ±0,5 °C (models with heated lid: ±0,3 °C)
  • Microprocessor controlled with temperature calibration via external thermometer or temperature device
  • Timer range: 1
More Product Information
Bestill nå

SPESIFIKASJONER

Environmentally Preferable Waste Reducing
B×D×H 127×160×123 mm 127×160×123 mm
Vekt 1,3 kg 1,3 kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR