VWR®, Block Heaters, Analogue
Leverandør: VWR Collection
460-0354EA 4750 NOK
460-0354 460-0355 460-0356 460-0357
VWR®, Block Heaters, Analogue
Termostatblokker Analoge termostatblokker
Multi-purpose, high wattage, constant temperature, VWR® analogue block heaters are economical, versatile and compact.
  • Compact design with small footprint
  • Temperature range, low: From ambient +5 to 100 °C, high: 75 to 150 °C
  • Multi-purpose device for a wide range of applications
  • Exceptional temperature uniformity
  • Accommodates VWR® interchangeable, modular heating blocks for tubes from 0,2 up to 50 ml capacity
  • Can be used at 18 to 33 °C, with 20 to 80% relative humidity, non condensing
These multi-purpose units are ideal for incubation and activationMore Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable Sustainable Packaging