Digital dry block heaters
Leverandør: IKA
4025100 4025200 4025300 4025400 4025102 4025302 4025402
460-1004EA 15900 NOK
460-1004 460-1005 460-1006 460-1007 460-0248 460-0261 460-0263
Digital dry block heaters
Termostatblokker Digitale termostatblokker
Digital block heaters which hold from one up to four aluminum blocks, provide precise temperature control in small vessels. The PT1000 temperature sensor allows temperature control directly in the sample vessel. It is used with PCR tubes, PCR strips, Greiner tubes, microplates and cuvettes. The block heaters are ideal for melting and boiling point determination, enzyme reactions, incubation and activation of cultures, immunoassays, DNA denaturation, culture media tests, coagulation tests, blood/urea nitrogen determinations and in situ hybridisations.
  • Stepless adjustable temperature