VWR® Heating Blocks for Block Heaters
Leverandør: VWR Collection
460-3236EA 1775 NOK
460-3236 460-3237 460-3238 460-3239 460-3224 460-3225 460-3226
VWR® Heating Blocks for Block Heaters
Termostatblokker Tilbehør til termostatblokker
Blokk: For bruk som lavtemperaturvarmeplate eller for spesialboring.
  • For use as a low-temperature hotplate or for custom drilling
Blocks for microtitre plates: Recessed well for better stability; flat surface is good for flat and round bottom plates. Fits 2/4/6 block heaters.
Blocks for PCR - plates, tubes, strips: Blocks for PCR - plates, tubes, strips: Tapered tube wells for 0,2 ml tubes. Spaced for easy access and removal.
Blocks for cuvettes: Two parallel slots fit six cuvettes in each slot, side by side for excellent stability and heat transfer.