VWR®, Accessories for Block Heaters Analogue/Digital
Leverandør: VWR Collection
460-3240EA 2150 NOK
460-3240 460-3241 460-3242 460-3289
VWR®, Accessories for Block Heaters Analogue/Digital
Termostatblokker Analoge termostatblokker
Plexiglas® covers with 2 open sides reduce airflow, providing additional temperature stability in low temperature applications.