PCR UV Cabinet - DNA/RNA, double PCR Workstation, UVT-S-AR
Leverandør: GRANT INSTRUMENTS
UVT-S-AREURO UVT-S-ARUK
461-0826EA 71000 NOK
461-0826 GRANUVT-S-ARUK
PCR UV Cabinet - DNA/RNA, double PCR Workstation, UVT-S-AR
Avtrekksskap og kabinetter PCR-arbeidsstasjoner og -skap
Advanced benchtop UV cabinet providing aseptic conditions for a variety of biomedical and biochemical procedures. The large capacity stainless steel UV cabinet with additional space for equipment and accessories allows more comfortable and convenient working in PCR working. With dual UV lamp protection.
  • Robust construction with large 1,2×0,52 m working area
  • UV surface irradiation – via single 25 W 254 nm open UV lamp
  • Patented built-in UV cleaner - recirculator with 25 m³/hour continuous air flow, prevents unwanted contamination and protects the user from