4,4'-Methylenedi-o-toluidine, Supelco®

Leverandør: Merck

Synonymer: 4,4'-Methylenebis(2-methylaniline)

MQ100

46106-100MG
46106-100MGEA 635 NOK
46106-100MG
4,4'-Methylenedi-o-toluidine, Supelco®
4,4'-Methylenedi-o-toluidine
Formel: C₁₅H₁₈N₂
MW: 226.3200 g/mol
MDL nummer: MFCD00126963
CAS-nummer: 838-88-0
EINECS: 212-658-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea534db25d2b668e3b87ae0684d9b91d00656dc2&vid=5f1c448060624ccfffb0a628754db1d1387cc7d9&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR