4-Aminoazobenzene, Supelco®

Leverandør: Merck

Synonymer: 4-Aminoazobenzene, p-Aminoazobenzene, 4-(Phenylazo)aniline, 4-Phenylazoaniline

M-Clarity™ quality level = MQ100

46130-250MG-R
46130-250MG-REA 490 NOK
46130-250MG-R
4-Aminoazobenzene, Supelco®
4-Aminoazobenzene
Formel: C₁₂H₁₁N₃
MW: 197.24 g/mol
MDL nummer: MFCD00007854
CAS-nummer: 60-09-3
EINECS: 200-453-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2d875b2a860076cfbd9c42326508170f1a67f524&vid=b8bd6c188dd9f6ec56e2d2b3a7b81dfe84e77cde&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR