Immersion coolers, Clifton, DC1-300
Leverandør: NICKEL ELECTRO
DC1-300
462-0021EA 57375 NOK
462-0021
Immersion coolers, Clifton, DC1-300
Termostatbad Termostatbad, lukket Termostatbad med kjøling
Immersion cooler without temperature control designed to be used in conjunction with Clifton NE4 stirred or circulating water baths, the unit runs continuously with the bath providing the temperature control. The immersion coil fits into the rear access hole behind the thermostirrer without loss of working area.
  • Bench top model, with 900 mm flexible hose, which is quiet in operation
  • Alternative to cooling with dry ice or tap water
  • Anti-bacterial paint finish that actively inhibits bacterial growth
Note: Cooling capacity at −20; −10; 0; 5 and 20 °C