Shaking water baths, Clifton NE5 Series
Leverandør: NICKEL ELECTRO
FC0025 FC0050 FC0100 FC0250 FC0500 FC1000 FC2000 NE5-28D NE5-10D NE5-28 SA01773
462-0022EA 340 NOK
462-0022 462-0023 462-0024 462-0025 462-0026 462-0027 462-0028 462-0050 462-0119 462-0118 452-0018
Shaking water baths, Clifton NE5 Series
Bad Ristevannbad
Digital and analogue linear shaking water baths with a stainless steel tank housed in a durable outer case. The corrosion resistant reciprocating trolley readily accepts perforated clip trays for holding flasks and test tube racks. The trolley is easily removed for cleaning purposes and a stainless steel recessed top accommodates flasks, clip trays etc prior to and after immersion. The 28 litre baths include a drain tap for convenient emptying. Digital shakers have splash proof controls featuring an advanced PID temperature controller with a LED display, allowing observation of set andMore Product Information