VWR®, Tube Racks for VWB Series Baths
Leverandør: VWR Collection
462-0253EA 1550 NOK
462-0253 462-0254 462-0255 462-0256 462-0257 462-0258 462-0252
VWR®, Tube Racks for VWB Series Baths
Stativer for rør Tube Racks
Tube racks suitable for: VWB 6, VWB 12, VWB 18, VWB 26, maximum 1, 4, 7, 7 racks per bath respectively.