VWR®, Tubing, Viton®, for Circulators
Leverandør: VWR Collection
462-0300EA 8325 NOK
462-0300
VWR®, Tubing, Viton®, for Circulators
Slanger Overføringsslange
Fluoroelastomer, black.
  • Excellent resistance to corrosive chemicals, oils, fuels and solvents
  • Resistant to ozone, sunlight and weathering
  • Tear resistant and durable
Temperature resistance: −40 to +150 °C.