Water baths, unstirred, SUB Aqua Pro series
Leverandør: GRANT INSTRUMENTS
SAP12 SAP18 SAP2 SAP26 SAP2S SAP34 SAP5 SAPD
462-0477EA 15000 NOK
462-0477 462-0478 462-0474 462-0479 462-0475 462-0480 462-0476 462-0481
Water baths, unstirred, SUB Aqua Pro series
Bad Vannbad, uten sirkulasjon
High quality, digital unstirred water bath range with choice of models from 2 to 34 litres and a dual bath with 5 and 12 litres. Dual baths are ideal when two different temperatures are needed and space is limited. All units have stainless steel tanks and durable painted steel cases, offer fast heat-up times, excellent temperature control, easy to use controls and a bright, clear display. All baths have heater mat technology ensuring optimum working space, without a heating element in the tank they are easier to clean. The 12 litre and larger models feature a convenient drain tap on theMore Product Information