Tissue float baths, Hydro H 2 P
Leverandør: LAUDA
L003071
462-0624EA 7350 NOK
462-0624
Tissue float baths, Hydro H 2 P
Bad Vevsbad
Tissue float bath for stretching and drying cut tissue samples. Working temperature range of the histology bath is 25 to 80 °C.
  • Temperature displayed on control thermometer at the bath edge
  • Temperature adjustment via rotary knob with temperature scale
  • 1-point temperature controller
  • Low water level protection
  • Black anodised aluminium bath interior
  • Control thermometer with holder as standard
Heater power: Max. 0,3 kW