Ion exchange resins, beads, BAKER ANALYZED® IONAC® NM-60 H+/OHˉ form, type I 16-50 netting (mixed bed exchanger), J.T.Baker®
Leverandør: Avantor
4631-01EA 2525 NOK
4631-01
Ion exchange resins, beads, BAKER ANALYZED® IONAC® NM-60 H+/OHˉ form, type I 16-50 netting (mixed bed exchanger), J.T.Baker®
Ion exchange resins
Sterk syre / sterk base, sulfonert/alkyl kvartær ammonium-polystyren
Bestill nå
Spesifikasjon Testresultater
Manufactured by IONAC® Chemical Company.
STORAGE: Protect from freezing and below 38°C (100°F)