Supelco® PESTANAL® Organic Reference Standard, Cycloheximide

Leverandør: Merck

46401-100MG-R
46401-100MG-REA 685 NOK
46401-100MG-R
Supelco® PESTANAL® Organic Reference Standard, Cycloheximide
Standarder Organiske standarder

  • GHS signal: GHS06, GHS08, GHS09


NB: P202 - P264 - P270 - P273 - P280 - P301 + P310

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=54d79286b388d76ee20e5b0ea99d83ed30ddd755&vid=54d79286b388d76ee20e5b0ea99d83ed30ddd755&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Synonym(s) 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide; Actidione; Naramycin A
Environmentally Preferable
M-Clarity quality level MQ100
CAS-nr 66-81-9
MDL No. MFCD00082346
Molekylvekt 281,35
Hardhetsklasse beskrivelse H300 - H341 - H360D - H411
Fraktforhold WICE
Lagringsforhold –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR