Supelco® Organic Reference Standard, rac-β-Tocopherol solution

Leverandør: Merck

46401-U
46401-UEA 3800 NOK
46401-U
Supelco® Organic Reference Standard, rac-β-Tocopherol solution
Standarder Organiske standarder

  • GHS signal: GHS02, GHS08, GHS07, GHS09


NB: P201 - P210 - P273 - P301 + P310 + P331 - P302 + P352 - P308 + P313

Bestill nå

SPESIFIKASJONER

M-Clarity quality level MQ100
CAS-nr 148-03-8
Hardhetsklasse beskrivelse H225 - H304 - H315 - H336 - H361f - H373 - H411
Flammepunkt –23 °C
Fraktforhold STD
Lagringsforhold –10...–25 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR