Monensin monosodium salt hydrate, Supelco®

Leverandør: Merck


M-Clarity™ quality level = MQ200

46468-100MG
46468-100MGEA 770 NOK
46468-100MG
Monensin monosodium salt hydrate, Supelco®
Monensin monosodium salt hydrate
Formel: C₃₆H₆₁NaO₁₁·H₂O
MW: 692.85 g/mol (anhy)
MDL nummer: MFCD00077826
CAS-nummer: 22373-78-0
EINECS: 244-941-7
FN: 3462
ADR: 6.1 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0397054656af8d165ea9b4e2489e570ecebda9ff&vid=f999573dd77dc8d93a85776544afe483a9c17d63&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR