EDTA dinatrium salt concentrate for dilution to 0.1 M in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®

Leverandør: Avantor

Synonymer: Idranal III, Edetic acid disodium salt, Kestranal® 2S, Ethylenedinitrilotetra-acetic acid disodium salt, Disodium dihydrogen ethylenediaminetetraacetate, Ethylenediaminetetra-acetic acid disodium salt

4653.EA 590 NOK
4653.
EDTA dinatrium salt concentrate for dilution to 0.1 M in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
EDTA dinatrium salt

(1/10 mole=33.62 g C₁₀H₁₄N₂Na₂O₈)

Formel: C₁₀H₁₄N₂Na₂O₈
MW: 336.21 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00070672
CAS-nummer: 139-33-3
EINECS: 205-358-3
Merck Indeks: 12,03556

Bestill nå

Spesifikasjon Testresultater

For Laboratory,Research,or Manufacturing Use
Molarity (by titrimetry) Passes Test
(Makes 0.1M solution after dilution to 1000 mL)

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR