Vacuum ovens, VO series
Leverandør: MEMMERT
VO29 VO49 VO101
466-0522EA 0 NOK
466-0522 466-0531 466-0543
Vacuum ovens, VO series
Ovner Vakuumovner
Reliable, powerful, energy efficient vacuum ovens with separately adjustable thermo shelves which are directly heated ensuring precise temperature control and, thanks to digital pressure control, the chamber load is not only dried at high speed, but also extremely gently in these robust units. Ideal for drying foodstuffs, cosmetics, granulates, watches, books, printed circuit boards, electronic components or injection moulds. All models have a safety glass door with bullet proof inner glass and an anti-splinter external screen. The exterior housing is made of textured stainless steel whileMore Product Information