Drying and Heating Chambers, with Forced Convection, M Avantgarde.Line
Leverandør: Binder
9010-0372 9010-0371 9010-0373 9010-0374
466-0702EA 0 NOK
466-0702 466-0701 466-0703 466-0704
Drying and Heating Chambers, with Forced Convection, M Avantgarde.Line
Testkammer Temperaturtestkammer
The M Series Avantgarde.Line units have a maximum of 300 °C and various advanced programming options ensure these material test chambers are ideal for material and aging tests. The units have a high air flow and their powerful fans ensure rapid heating.
  • Temperature range: 15 to 22 °C above ambient temperature up to +300 °C
  • High temperature accuracy thanks to APT.line™ technology with increased heating output
  • Reinforced fan with adjustable speed
  • Electromechanical control of the exhaust air flap
  • Intuitive touchscreen controller with time-segment and