Drying and Heating Chambers, with Forced Convection, M Avantgarde.Line
Leverandør: Binder
9010-0372 9010-0371 9010-0373 9010-0374
466-0702EA 0 NOK
466-0702 466-0701 466-0703 466-0704
Drying and Heating Chambers, with Forced Convection, M Avantgarde.Line
Testkammer Temperaturtestkammer
The M Series Avantgarde.Line units have a maximum of 300 °C and various advanced programming options ensure these material test chambers are ideal for material and aging tests. The units have a high air flow and their powerful fans ensure rapid heating.
  • Temperature range: 15 to 22 °C above ambient temperature up to +300 °C
  • High temperature accuracy thanks to APT.line™ technology with increased heating output
  • Reinforced fan with adjustable speed
  • Electromechanical control of the exhaust air flap
  • Intuitive touchscreen controller with time-segment and
More Product Information
Bestill nå
SPESIFIKASJONER
Modell M 115 M 56 M 260 M 720
Temperaturområde Ambient 22…300 °C Ambient 22…300 °C Ambient 22…300 °C Ambient 22…300 °C
Environmentally Preferable
Temperatursvingning (tid) ±0,2 K @ 150 °C ±0,2 K @ 150 °C ±0,3 K @ 150 °C ±0,5 K @ 150 °C
Temperaturvariasjon (romlig) ±1,4 K @ 150 °C ±1,4 K @ 150 °C ±1,7 K @ 150 °C ±2 K @ 150 °C
Kapasitet 113 L 59 L 256 L 735 L
Konveksjontypen tvungen konveksjon tvungen konveksjon tvungen konveksjon tvungen konveksjon
Innvendig B×D×H 550×375×550 mm 400×335×440 mm 650×505×780 mm 1000×565×1300 mm
Utvendig B×D×H 710×636×732 mm 560×596×622 mm 810×786×962 mm 1160×816×1590 mm
Maks. belastning per hylle 30 kg/shelf; 150 kg/unit 30 kg/shelf; 70 kg/unit 40 kg/shelf; 270 kg/unit 45 kg/shelf; 315 kg/unit
Strømforbruk 2,4 kW 2 kW 3,1 kW 6 kW
Gjenvinningstid 7 min after 30 s opening at 150 °C 4 min after 30 s opening at 150 °C 7 min after 30 s opening at 150 °C 7 min after 30 s opening at 150 °C
Tilleggshyller (max.) 2 (4) 2 (4) 2 (8) 2 (16)
Vekt 56 kg 43 kg 85 kg 169 kg