Argatroban

Leverandør: Thermo Fisher Scientific

Synonymer: MCI-9038

466390010 466392500
466390010.EA 14350 NOK
466390010. 466392500.
Argatroban
Argatroban
Formel: C₂₃H₃₆N₆O₅S
MDL nummer: MFCD23102419
CAS-nummer: 74863-84-6

Bestill nå

Spesifikasjon Testresultater

Water =<4 %
Appearance (Form) Powder
HPLC >=97.5 %
Appearance (Color) White to almost white
Infrared spectrum Conforms


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR