Eksikatorskap, SICCO Auto Star
Leverandør: Bohlender
V1877-07
467-0054EA 37500 NOK
467-0054
Eksikatorskap, SICCO Auto Star
Eksikatorer Eksikatorskap
This transparent acrylic cabinet has a reinforced aluminium frame, door with foam rubber gasket and magnetic strip. Suitable for long-term storage, frequent opening of the door is compensated automatically. Ideal solution for storing reference samples, electronics, photographic equipment, papers and valuable cultural assets.
  • Door mounted electronic hygrometer indicates minimum and maximum temperature and humidity values
  • Automatic drying which enables constant low interior humidity between 20 and 30%
  • Temperature resistance from −20 to +70 °C
  • Cabinet has non