Mini desiccator cabinets, SICCO, Black
Leverandør: Bohlender
V1840-06 V1940-06
467-0124EA 4275 NOK
467-0124 467-0127
Mini desiccator cabinets, SICCO, Black
Eksikatorer Eksikatorskap
Opaque black polycarbonate panels reduce light incidence and protect samples from ultraviolet radiation. Door with snap fit closing and integrated gasket ensuring easy opening and tight closure. Temperature resistant from −35 to +70 °C, suitable for use under normal pressure.
  • Large opening and door with wide opening angle enable user to access the entire interior space
  • Robust construction, lightweight, portable and stackable
  • Premium model includes hygrometer and two shelves
Leveringsinformasjon: Supplied with desiccant which is in a handy, resealable container.