Kaliumdikromat concentrate for dilution to 1/60 mol in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®

Leverandør: Avantor

4671.EA 330 NOK
4671.
Kaliumdikromat concentrate for dilution to 1/60 mol in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Kaliumdikromat
Formel: K₂Cr₂O₇
MW: 294.18 g/mol
Kokepunkt: 500
Smeltepunkt: 398
MDL nummer: MFCD00011367
CAS-nummer: 7778-50-9
FN: 3087
ADR: 5.1 II
Merck Indeks: 12,07790

Bestill nå

Spesifikasjon Testresultater

Titer (N) 0.0998-0.1002


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR