Topotecan hydrochloride hydrate 97%

Leverandør: Thermo Fisher Scientific

467180010 467182500
467180010.EA 6450 NOK
467180010. 467182500.
Topotecan hydrochloride hydrate 97%
Topotecan hydrochloride hydrate
Formel: C₂₃H₂₃N₃O₅·xHCl·yH₂O
MDL nummer: MFCD28989044
CAS-nummer: 119413-54-6

Bestill nå

Spesifikasjon Testresultater

Appearance yellow crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %
Loss on drying ≤14 %
Titration Argentometric ≤17.3 % (as HCl)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR