Kaliumjodat concentrate for dilution to 1/60 mol/l (0.1 N) in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®

Leverandør: Avantor

4676.EA 960 NOK
4676.
Kaliumjodat concentrate for dilution to 1/60 mol/l (0.1 N) in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Kaliumjodat
Formel: KlO₃
MW: 214 g/mol
Smeltepunkt: 560
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011406
CAS-nummer: 7758-05-6
EINECS: 231-831-9
Merck Indeks: 13,07726

Bestill nå

Spesifikasjon Testresultater

For Laboratory,Research,or Manufacturing Use 0.0998-0.1002
Normality (by titrimetry) Passes Test
(Makes 0.1N solution after dilution to 1000 mL)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR